VWR®, Spreaders, Triangle Shaped, Disposable
Lieferant: VWR Collection
Zertifikate
Über diesen Artikel
PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial culture work. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
- Durable enough to be autoclaved and used again
- Width: 30 mm (blue)/60 mm (yellow)
Zugehörige Informationen
Frequently Bought Together