Order Entry
Austria
ContactUsLinkComponent
Biohazard transport carriers, Nalgene®
Biohazard transport carriers, Nalgene®
Catalog # 113-8185
   Low Manufacturing Impact
undefined
Biohazard transport carriers, Nalgene®
Catalog # 113-8185
   Low Manufacturing Impact

Specifications

  • Description:
    Test tube carrier
  • W×D×H:
    184×368×171 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1

Specifications

About this item

Transparent PC with silicone gasket and PC clamps.

  • For 13 or 16 mm Vacutainer test tubes
  • Adaptable for other test tubes
  • Tough and break resistant with excellent visibility of contents
  • Easy to grasp side handles are moulded in
  • Clamps hold the carrier securely closed and ensure leakproof seal

This closed system carrier is designed to protect the lab worker during transportation of tubes filled with potentially hazardous samples. It is autoclavable without removing lid or opening clamps to avoid hazardous exposure. Warning labels are included.

Meets OSHA Standard 29 CFR Part 1910.1030 for use as protection against blood borne pathogens.

Certifications: Meets OSHA Standard 29 CFR Part 1910.1030 for use as protection against blood borne pathogens.
Ordering information: Stainless steel handle to be ordered separately.