Desktop Calculators
Desktop calculator HS-12 ER, 12 digit, 151×152×32 mm, 181 g
818-3420
Solar powered 12-digit, angled LCD with additional support, to improve the reading angle.
- Quick correction button
- Sign change button
- Automatic rounding
Functions include: € conversion - percentage/delta percentage calculation - floating and fixed-point calculation - subdivision to the thousand - full/constant memory.
: Includes batteries.
- Weight:181 g
- Description:Desktop calculator HS-12 ER, 12 digit
- W×D×H:151×152×32 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



