Order Entry
Austria
ContactUsLinkComponent
Connection Cable for Non-Heating Apparatus
Connection Cable for Non-Heating Apparatus
Catalog # 139-1379
Supplier:  BACHMANN ELEKTRO
undefined
Connection Cable for Non-Heating Apparatus
Catalog # 139-1379
Supplier:  BACHMANN ELEKTRO

Specifications

  • Colour:
    Grey
  • Description:
    IEC lead cable
  • Length:
    3 m
  • Pk:
    1

Specifications

About this item

Supply line for earthed devices 65 °C.

  • With protective angled plug und earthed appliance socket

Colour: grey

Certifications: VDE-compliant