Orders LinkContactUsLinkComponent
Automatic burettes, pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
 :   Brand
Automatic burettes, pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
 :  Brand
Automatic burettes, pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
Automatic burettes, pellet pattern, without intermediate stopcock, Schellbach, BLAUBRAND®
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Automatic zeroing, needle-valve stopcock with PTFE spindle, without intermediate stopcock
DIN EN ISO 385
 : DIN EN ISO 385
 : Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
Product Family Options
Automatic burette supplied without reservoir bottle
  • Capacity
    Division
    Total height
    Tolerance
  • 10 ml
    0,02 ml
    1000 mm
    0,02 ± ml
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
  • 25 ml
    0,05 ml
    1000 mm
    0,03 ± ml
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
  • 50 ml
    0,1 ml
    1000 mm
    0,05 ± ml
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
Accessories
  •  
    Description
    Capacity
  • 612-9953
    Ball set with net, black, size 5, natural rubber/PVC
    102
    1 * 1 ST - 107,00 €
  •  
    Description
    Capacity
  • 612-3219
    Reservoir bottle, clear glass
    2000 ml
    1 * 1 ST - 74,30 €