Burettes, straight stopcock, Azlon®
Supplier: SciLabware
Certificates
About this item
Acrylic, with PMP/PE stopcocks, class B.
- Graduations are screen printed to ISO 385 class B accuracy
- Unaffected by most dilute aqueous solutions (except ammonia and hydrofluoric acid) but unsuitable for use with ethanol or organic solvents
- Suitable for both left and right handed use
Acrylic construction for strength and transparency. Suitable for use in education, research or industrial laboratories, especially where breakages often occur.
Delivery: Supplied with a technical data sheet.
Frequently Bought Together