Burettes, lateral stopcock, Schellbach
DURAN®, borosilicate glass, class AS.
- Calibrated to deliver 'Ex', conformity certified (DE-M marking) with dated batch identification
- Schellbach stripe, ring marks at major graduations
- Blue graduations
Scale length: 500 mm
Scale length: 500 mm
: DIN EN ISO 385
Frequently Bought Together