Orders LinkContactUsLinkComponent
Buckets
Supplier:   VITLAB
Buckets
Supplier:  VITLAB
Certificates
Buckets
4 Options Available Below
Product Details & Documents
HDPE, white.
  • Good chemical and temperature resistance
  • Graduated with litre scale
  • Without spout
For industrial and laboratory applications. With handle and tight closing, transparent lid.
Product Family Options
With lid
Product Information
  • Colour
    Capacity
    Ø ext.
    Height
    Availability
    Price
  • White
    5 L
    235 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €
  • White
    10 L
    290 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,30 €
    24,30 €
    1 * 1 ST
       24,30 €
Without lid
Product Information
  • Colour
    Capacity
    Ø ext.
    Height
    Availability
    Price
  • White
    5 L
    250 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,30 €
    16,30 €
    1 * 1 ST
       16,30 €
  • White
    10 L
    290 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,50 €
    19,50 €
    1 * 1 ST
       19,50 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lid, PP, for 10 l bucket
    Lid, PP, for 10 l bucket
    216-1505
    Log in for availability
    1 * 1 ST - 5,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Lid, PP, for 5 l bucket
    Lid, PP, for 5 l bucket
    216-1504
    Log in for availability
    1 * 1 ST - 4,60 €