Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Slide boxes
Certificates
Slide boxes
4 Options Available Below
Product Details & Documents
These slide boxes are designed for secure, dust-free storage of specimen slides with metal catch and hinges.
  • Numbered index in the lid
  • Cardboard with black wooden frame
  • Numbered plastic grooves: Anti-static and xylene-resistant
Suitable for the safe and protected storage of slides.
Documents
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    Availability
    Price
  • 631-0353
    Slide box for micro slides
    Slide box for micro slides
    Black/cream
    Wood/cardboard
    25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,40 €
    17,40 €
    1 * 1 ST
       17,40 €
  • 631-0352
    Slide box for micro slides
    Slide box for micro slides
    Black/cream
    Wood/cardboard
    50
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • HECH42701020
    Slide box for micro slides
    Slide box for micro slides
    Black/cream
    PP
    100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • HECH42701010
    Slide box for micro slides
    Slide box for micro slides
    Black/cream
    Wood/cardboard
    100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,50 €
    18,50 €
    1 * 1 ST
       18,50 €