Orders LinkContactUsLinkComponent
Cool Boxes
Cool Boxes
Supplier:   CAMPING GAZ
Cool Boxes
Supplier:  CAMPING GAZ
Certificates
Cool Boxes
2 Options Available Below
Product Details & Documents
Blue-white PP. Sturdy, robust design, ideal for storing and transporting temperature-sensitive materials and samples.
  • Box casing and lid fully insulated with CFC-free PU foam
  • A rubber seal ensures the boxes are airtight, a lid with non breakable handle can be securely locked
  • Excellent cooling performance (depending on the model up to 24 hours cooling)
Product Family Options
Product Information
  • Capacity
    Dimensions
    Availability
    Price
  • 216-6985
    Cooling box ISOTHERM Extreme
    Cooling box ISOTHERM Extreme
    10 l
    310×225×350 mm (ext); 265×165×270 mm (int)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,10 €
    49,10 €
    1 * 1 ST
       49,10 €
  • 216-6981
    Cooling box ISOTHERM Extreme
    Cooling box ISOTHERM Extreme
    17 l
    365×295×450 mm (ext); 320×195×310 mm (int)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,80 €
    56,80 €
    1 * 1 ST
       56,80 €