Bottles, wide neck, square, with screw cap, Azlon®
Supplier: SciLabware
Certificates
About this item
HDPE, translucent.
- Heavy-duty bottles with plug seal and screw cap
- Easy to grip caps have a toggle for tamper-evident wire attachment to neck
- Moulded graduations for approximate volume measurement, food safe
Frequently Bought Together