Bottles, wide neck, with screw cap, Mason type, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, translucent with PP screw cap.
- Very good chemical resistance
- Heat resistant up to 121 °C, autoclavable
- Leakproof with ring for tamper evident heat shrink bands
Bottles have 70 mm necks that accept fixtures or closures with Mason jar threads. Also fit on mixers equipped with the special Mason thread.
Specifications
- Catalog No:
- 215-7851
- 215-7852
- 215-7853
- 215-7854
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement