Wash bottles, narrow neck, autoclavable, Nalgene®
PPCO, translucent, PP spray cap.
- Very good chemical resistance, heat resistant up to +121 °C
- Flexible, leakproof closure
- With blue 'Autoclavable' label and printed size code
- Catalog No:
- 215-8107
- 215-8108
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together