Wash bottles, integral, narrow neck, Azlon®
Supplier: SciLabware
Certificates
About this item
LDPE, translucent, with PP spray cap.
- Slim hexagonal cross section makes the bottles particularly suited to smaller hands
- Precision tip is ideal for use as a dropper or for fine liquid jet dispensing
Ergonomic wash bottles with an integral moulded dispensing tube and removable jet tip.
Frequently Bought Together