Order Entry
Austria
Orders LinkContactUsLinkComponent
Sample bottles

Sample bottles

Supplier:   GRAVIS
Sample bottles
Supplier:  GRAVIS
Certificates
Sample bottles

2 Options Available Below

Product Details & Documents

Glass.

  • Screw ring
  • Different sizes
  • Capsules of different formats and materials for a wide variety of applications

Wide neck.

PH30 and PH40 ring.

Delivery information: Supplied without capsules (to be selected separately).

Product Family Options

Round bottles, threaded wide neck, without capsules

Product Information

  • Capacity
    Thread
    Ext.Ø
    Height
    Neck I-Ø
    Availability
    Price
  • 100 ml
    40 mm
    49,4 mm
    87,2 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 86 ST
    1 * 86 ST - 122,00 €
    122,00 €
    1 * 86 ST
       122,00 €

  • 120 ml
    40 mm
    48,5 mm
    106,1 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 86 ST
    1 * 86 ST - 145,00 €
    145,00 €
    1 * 86 ST
       145,00 €

Screw caps, Bakelite

Product Information

  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    40 mm
    with TRISEAL seal
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 82,80 €
    82,80 €
    1 * 100 ST
       82,80 €

Product Information

  • Catalog #
    Thread
    Description
    Colour
    Availability
    Price
  • 07
    with Butyl/PTFE seal
    40 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 358,00 €
    358,00 €
    1 * 100 ST
       358,00 €

Product Information

  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    30 mm
    with rubber liner
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,50 €
    90,50 €
    1 * 100 ST
       90,50 €

Product Information

  • Catalog #
    Thread
    Description
    Colour
    Availability
    Price
  • 05
    with Butyl/PTFE seal
    30 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 248,00 €
    248,00 €
    1 * 100 ST
       248,00 €

Product Information

  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    40 mm
    with rubber liner
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 141,00 €
    141,00 €
    1 * 100 ST
       141,00 €

Product Information

  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    30 mm
    with TRISEAL seal
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 57,70 €
    57,70 €
    1 * 100 ST
       57,70 €

Screw caps, Aluminium

Product Information

  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Silver
    with rubber liner
    40 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 124,00 €
    124,00 €
    1 * 100 ST
       124,00 €

Product Information

  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Silver
    with rubber liner
    30 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 76,90 €
    76,90 €
    1 * 100 ST
       76,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.