Bottles, wide neck, with standard ground stoppers
Soda-lime glass, clear or brown, with standard ground joint and glass stopper.
- Good chemical and physical properties
- For use in applications that are not associated with high thermal loads
- Stable and tough
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
: DIN 12039
Frequently Bought Together