Bottles, rectangular, narrow neck with screw caps, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
HDPE, translucent, graduated with red tamper evident screw caps.
- Rectangular cross section bottles save storage space
- Graduated bottles are supplied with caps but are unassembled
- Sterile bottles are irradiated and are supplied capped
These bottles are graduated and sterile available. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.
Specifications
- Catalog No:
- 215-0702
- 215-0700
- 215-0699
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre