Bottles, narrow neck, square, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent or PPCO, translucent, with PP screw cap.
- HDPE: Good chemical resistance/PPCO: Very good chemical resistance
- HDPE: Heat resistant up to 120 °C/PPCO: Heat resistant up to 135 °C, autoclavable
- Leakproof
Ideal for storing and shipping liquids. Graduated.
Specifications
- Catalog No:
- 215-7641
- 215-7621
- 215-7622
- 215-7646
- 215-7625
- 215-7644
- 215-7645
- 215-7642
- 215-7626
- 215-7643
- 215-7623
- 215-7624
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ACT Label
- NAR Certificate of Retirement
- NAR Certificate of Retirement