Bottles, narrow neck, rectangular, LDPE, with thread
About this item
LDPE, translucent, without screw cap. Ideal for storing and shipping liquids. Graduated.
- Good chemical resistance
- Heat resistant up to +80 °C
- Side ribbing for better grip
Bestellinformation: The bottles are supplied without caps. For suitable caps for 125/500 and 1000 ml please see 215-9226 (for 250 ml botlles see MUEL14.063.01).
4 Options Available Below
Product Details & Documents
LDPE, translucent, without screw cap. Ideal for storing and shipping liquids. Graduated.
- Good chemical resistance
- Heat resistant up to +80 °C
- Side ribbing for better grip
Bestellinformation: The bottles are supplied without caps. For suitable caps for 125/500 and 1000 ml please see 215-9226 (for 250 ml botlles see MUEL14.063.01).
Product Family Options
Product Information