Orders LinkContactUsLinkComponent
Jars, with twist-off thread
Jars, with twist-off thread
Jars, with twist-off thread
Supplier:  MUELLER KREMPEL
Certificates
Jars, with twist-off thread
Jars, with twist-off thread
6 Options Available Below
Product Details & Documents
Amber glass, without lid.
  • Good chemical and physical properties
  • For uses without high thermal load
  • Matching metal lid available as an separately (see 216-3661 and 216-3663)
Well suited for sampling, storage and transport of powders, pastes and liquids. Amber coloured to protect light-sensitive substances.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Thread
    Availability
    Price
  • 60 ml
    50,0 mm
    68,0 mm
    43 mm
    Log in for availability
    Log in for availability
    1 * 99 ST
    1 * 99 ST - 102,00 €
    102,00 €
    1 * 99 ST
       102,00 €
  • 85 ml
    50,0 mm
    82,0 mm
    43 mm
    Log in for availability
    Log in for availability
    1 * 68 ST
    1 * 68 ST - 87,30 €
    87,30 €
    1 * 68 ST
       87,30 €
  • 125 ml
    50,0 mm
    104,0 mm
    43 mm
    Log in for availability
    Log in for availability
    1 * 56 ST
    1 * 56 ST - 86,50 €
    86,50 €
    1 * 56 ST
       86,50 €
  • 250 ml
    65,0 mm
    122,0 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 104,00 €
    104,00 €
    1 * 48 ST
       104,00 €
  • 630 ml
    85,0 mm
    163,0 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 99,60 €
    99,60 €
    1 * 20 ST
       99,60 €
  • 1000 ml
    100,0 mm
    179,0 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 14 ST
    1 * 14 ST - 78,00 €
    78,00 €
    1 * 14 ST
       78,00 €