ContactUsLinkComponent
Bottle heads, adjustable for gas washing bottles, Drechsel pattern, Quickfit®
Bottle heads, adjustable for gas washing bottles, Drechsel pattern, Quickfit®
Catalog # 201-8629
Supplier:  SciLabware
undefined
Bottle heads, adjustable for gas washing bottles, Drechsel pattern, Quickfit®
Catalog # 201-8629
Supplier:  SciLabware
Specifications
  • Thread:
    Size 13
  • Description:
    24/29 NS
  • Pk:
    1
Specifications
About this item
PYREX®, borosilicate glass, without glass filter disc.
  • Very good chemical resistance
  • High temperature resistance
With screwthread joint which allows height adjustment of the dip tube and hence permits use with either 125 ml or 250 ml Drechsel bottles. Detachable plastic screwthreads connectors for easy fitting of flexible tubing.