Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Supplier:   Lenz Laborglas
Dissolved oxygen bottles
Supplier:  Lenz Laborglas
Certificates
Dissolved oxygen bottles
2 Options Available Below
Product Details & Documents
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Product Family Options
Product Information
  • Capacity
    NS stopper
    Availability
    Price
  • 528-1203
    Oxygen bottles acc. to Winkler
    Oxygen bottles acc. to Winkler
    250-300 ml
    19/26
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,50 €
    34,50 €
    1 * 1 ST
       34,50 €
  • 528-1205
    Oxygen bottles acc. to Winkler
    Oxygen bottles acc. to Winkler
    100-150 ml
    14/23
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,90 €
    32,90 €
    1 * 1 ST
       32,90 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 14/23 for Dissolved Oxygen Bottles
    Clamps NS 14/23 for Dissolved Oxygen Bottles
    705-0564
    Log in for availability
    1 * 1 ST - 7,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 19/26 for Dissolved Oxygen Bottles
    Clamps NS 19/26 for Dissolved Oxygen Bottles
    705-0565
    Log in for availability
    1 * 1 ST - 8,20 €