ContactUsLinkComponent
Conical-bottom centrifuge tube adapters, Nalgene®
Conical-bottom centrifuge tube adapters, Nalgene®
Catalog # 525-0143
   Low Manufacturing Impact
undefined
Conical-bottom centrifuge tube adapters, Nalgene®
Catalog # 525-0143
   Low Manufacturing Impact
Specifications
  • Ø:
    28,83 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    6
Specifications
About this item
PC, autoclavable.
  • These adapters must be used with Nalgene® conical-bottom centrifuge tubes (35 ml)
  • Fits most commonly used low- and high-speed rotors
  • Easily removed from rotors after use