Orders LinkContactUsLinkComponent
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
45 Options Available Below
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Product Family Options
Thread size 8-425
Product Information
  • Thread
    Cap colour
    Availability
    Price
  • 8-425
    Pink
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4004
  • 8-425
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4005
  • 8-425
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4006
  • 8-425
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4007
  • 8-425
    Grey
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4008
  • 8-425
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4009
  • 8-425
    Purple
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4010
  • 8-425
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-2763
  • 8-425
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,30 €
    48,30 €
    1 * 100 ST
       48,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4011
  • 8-425
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 450,00 €
    450,00 €
    1 * 1.000 ST
       450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4037
  • 8-425
    Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 450,00 €
    450,00 €
    1 * 1.000 ST
       450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4038
  • 8-425
    Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 450,00 €
    450,00 €
    1 * 1.000 ST
       450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4039
  • 8-425
    Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 450,00 €
    450,00 €
    1 * 1.000 ST
       450,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4040
  • 8-425
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 440,00 €
    440,00 €
    1 * 1.000 ST
       440,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4041
Thread size 13-425
Product Information
  • Thread
    Cap colour
    Availability
    Price
  • 13-425
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4029
  • 13-425
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4030
  • 13-425
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4031
  • 13-425
    Grey
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,20 €
    55,20 €
    1 * 100 ST
       55,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4032
  • 13-425
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4033
  • 13-425
    Purple
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4034
  • 13-425
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 55,60 €
    55,60 €
    1 * 100 ST
       55,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4035
  • 13-425
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,20 €
    54,20 €
    1 * 100 ST
       54,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4036
Thread size 15-415
Product Information
  • Thread
    Cap colour
    Availability
    Price
  • 15-415
    Pink
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4012
  • 15-415
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-2764
  • 15-415
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4013
  • 15-415
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4014
  • 15-415
    Grey
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4015
  • 15-415
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4016
  • 15-415
    Purple
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4017
  • 15-415
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,20 €
    51,20 €
    1 * 100 ST
       51,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4018
  • 15-415
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 50,20 €
    50,20 €
    1 * 100 ST
       50,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4019
  • 15-415
    Red
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    WHEAW242512-A
  • 15-415
    Grey
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 551,00 €
    551,00 €
    1 * 1.000 ST
       551,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4042
  • 15-415
    White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 541,00 €
    541,00 €
    1 * 1.000 ST
       541,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4043
Thread size 20-410
Product Information
  • Thread
    Cap colour
    Availability
    Price
  • 20-410
    Pink
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4020
  • 20-410
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4021
  • 20-410
    Orange
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4022
  • 20-410
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-2765
  • 20-410
    Grey
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4023
  • 20-410
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4024
  • 20-410
    Purple
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4025
  • 20-410
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4026
  • 20-410
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,30 €
    63,30 €
    1 * 100 ST
       63,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4027
  • 20-410
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 571,00 €
    571,00 €
    1 * 1.000 ST
       571,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4044
  • 20-410
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 571,00 €
    571,00 €
    1 * 1.000 ST
       571,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    WHEAW242522-A
  • 20-410
    Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 571,00 €
    571,00 €
    1 * 1.000 ST
       571,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    215-4045