Orders LinkContactUsLinkComponent
Double bosshead
Aluminium alloy, powder coated, 13 mm
  241-0370
Double bosshead
  241-0370
Double bosshead
Double bosshead
Aluminium alloy, powder coated, 13 mm
13,60 €
1 * 1 ST
Double bosshead made from powder-coated aluminium alloy.
With nickel plated brass thumb screws.
  • Material:
    Aluminium alloy, powder coated
  • For rod Ø:
    13 mm