VWR® Bossheads
For assembling 12, 13 and 16 mm diameter rods.
- Bossheads can be fully opened; this enables the rod to be securely attached in the desired position
- Easy to assemble and take apart by means of an Allen key (optional, 241-0053)
- Excellent chemical resistance, robust
* When stainless steel boss is locked onto the stainless steel rod it cannot be moved by a rotational force less than 3,5 Nm.
Frequently Bought Together