Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
25 mm, Tempered cast iron
Catalog # 241-2550
Supplier:   Lenz Laborglas
Double bosshead
Catalog # 241-2550
Supplier:  Lenz Laborglas
Double bosshead
Double bosshead
25 mm, Tempered cast iron
Price
59,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Bosshead made from tempered cast iron. For attaching reaction vessel clamps to support stands.
  • Clasping range up to 25 mm
  • Fast and easy assembly and disassembly via large brass tommy screws
  • Robust, durable
 : DIN 12895
Specifications
  • Spanning width:
    25 mm
  • Material:
    Tempered cast iron