ContactUsLinkComponent
Filtering beakers, DURAN®
Filtering beakers, DURAN®
Catalog # 511-0320
Supplier:  DWK Life Sciences
Supplier #: 211316805
undefined
Filtering beakers, DURAN®
Catalog # 511-0320
Supplier:  DWK Life Sciences
Supplier #: 211316805
Log in for availability
Select an Option
157 mm
Select an Option
3000 ml
Select an Option
Graduated
Select an Option
205 mm
Select an Option
08
Select an Option
1
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Increased wall thickness for better mechanical properties
  • Limited use as cooking glass due to reduced thermal shock resistance
  • Available graduated or non graduated
Thick walled, with spout, easy to read scale and large labelling field.
Above 10000 ml without graduation
Frequently Bought Together