Orders LinkContactUsLinkComponent
Measuring beaker with handle
Measuring beaker with handle
Supplier:   NICKEL ELECTRO
Measuring beaker with handle
Supplier:  NICKEL ELECTRO
Certificates
Measuring beaker with handle
Measuring beaker with handle
3 Options Available Below
Product Details & Documents
Stainless steel, rust-proof.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for fast, easy cleaning
Conical, without rim, with broad base and textured graduations.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,70 €
    44,70 €
    1 * 1 ST
       44,70 €
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,00 €
    58,00 €
    1 * 1 ST
       58,00 €
  • 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €