ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # 213-0262
Supplier:  VITLAB
Supplier #: 441941
undefined
Measuring beakers with handle
Catalog # 213-0262
Supplier:  VITLAB
Supplier #: 441941
Log in for availability
Select an Option
92 mm
Select an Option
10 ml
Select an Option
500 ml
Select an Option
140 mm
Select an Option
04
Select an Option
1
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.
Frequently Bought Together