Order Entry
Austria
ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # 213-3745
Supplier:  VITLAB
Supplier #: 481941
undefined
Measuring beakers with handle
Catalog # 213-3745
Supplier:  VITLAB
Supplier #: 481941
Log in for availability
Select an Option
97 mm
Select an Option
10 ml
Select an Option
500 ml
Select an Option
140 mm
Select an Option
02
Select an Option
1

About this item

PP, highly translucent.

  • Volume is equally visible for left and right handers
  • Good chemical resistance, resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • To preserve markings, cleaning at no higher than +60 °C is recommended
  • For autoclaving we recommend the design with raised graduations

Ideal for general laboratory use and a wide variety of other applications. With stable handle, spout and easily readable, printed black scale on both sides. Stackable.

Certifications: Food-safe products acc. to EC Directive No. 10/2011.
Frequently Bought Together