ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # 213-5115
Supplier #: L51/8
undefined
Measuring beakers with handle
Catalog # 213-5115
Supplier #: L51/8
Log in for availability
Select an Option
110 mm
Select an Option
1000 ml
Select an Option
165 mm
Select an Option
03
Select an Option
1
About this item
Porcelain, glazed white except for the base.
  • Exceptional chemical resistance
  • Very good thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. With very easy-to-read, internal graduations. Label with the nominal volume on the outside of the measuring beaker.
Frequently Bought Together