Jacketed beakers with hose connections
Artikelnr. 201-0372
Supplier: NEUBERT VOLUME GLASSWAERE
Supplier #: 0710-0250-55
Log in for availability
About this item
Borosilicate glass 3.3, without spout.
- Very good chemical resistance
- Very good thermal resistance
- With glass tubing connectors O.Ø: 10 mm
For maintaining liquid samples at a constant temperature. The temperature controlling medium is pumped through the double-walled jacket by means of a recirculating thermostat.
Frequently Bought Together