Beakers, tall form, DURAN®
Catalog # 801-3136
Supplier: DWK Life Sciences
Supplier #: 211173603
Log in for availability
About this item
DURAN®, borosilicate glass 3.3, clear, without spout.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
With printed Retrace Code (batch identification).
Certifications: ISO 3819
Frequently Bought Together