Orders LinkContactUsLinkComponent
Beakers, low form | Avantor
Beakers, low form
Supplier:   KARTELL
Beakers, low form
Supplier:  KARTELL
Certificates
Beakers, low form
Beakers, low form
9 Options Available Below
Product Details & Documents
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use at 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
ISO 7056
Certifications: ISO 7056.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 25 ml
    49 mm
    34 mm
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,90 €
    4,90 €
    1 * 1 ST
       4,90 €
  • 50 ml
    60 mm
    41 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,10 €
    3,10 €
    1 * 1 ST
       3,10 €
  • 100 ml
    72 mm
    51 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,90 €
    4,90 €
    1 * 1 ST
       4,90 €
  • 250 ml
    95 mm
    71 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,80 €
    6,80 €
    1 * 1 ST
       6,80 €
  • 500 ml
    119 mm
    87 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,50 €
    9,50 €
    1 * 1 ST
       9,50 €
  • 1000 ml
    147 mm
    109 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,90 €
    16,90 €
    1 * 1 ST
       16,90 €
  • 2000 ml
    183 mm
    132 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,40 €
    22,40 €
    1 * 1 ST
       22,40 €
  • 3000 ml
    201 mm
    158 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,60 €
    35,60 €
    1 * 1 ST
       35,60 €
  • 5000 ml
    229 mm
    188 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,20 €
    54,20 €
    1 * 1 ST
       54,20 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.