ContactUsLinkComponent
VWR®, Beaker Cups, Disposable
VWR®, Beaker Cups, Disposable
Catalog # 611-1368
Supplier:  VWR Collection
Supplier #:
undefined
VWR®, Beaker Cups, Disposable
Catalog # 611-1368
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
250 ml
Select an Option
88 mm
Select an Option
07
Select an Option
100
About this item
PS.
  • Suitable for use in various sample dispensers
  • Beakers with capacity of 5 and 20 ml are recommended for use in flame photometric analyses
Economical, multipurpose beaker for a very wide range of uses.
Frequently Bought Together