Orders LinkContactUsLinkComponent
Beakers, low form, Azlon®
Beakers, low form, Azlon®
Beakers, low form, Azlon®
Supplier:  SciLabware
Certificates
Beakers, low form, Azlon®
Beakers, low form, Azlon®
6 Options Available Below
Product Details & Documents
PP, highly translucent.
  • Tapered stacking style - space saving and easy to handle
  • Autoclavable, durable with good chemical resistance
  • Precise non-drip pouring spouts, with easy-to-read moulded graduations
Certifications: Comply with ISO 7056.
Documents
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 50 ml
    61 mm
    39 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,70 €
    24,70 €
    1 * 10 ST
       24,70 €
  • 100 ml
    73 mm
    48,5 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 30,30 €
    30,30 €
    1 * 10 ST
       30,30 €
  • 250 ml
    96 mm
    65,5 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 38,50 €
    38,50 €
    1 * 10 ST
       38,50 €
  • 500 ml
    121 mm
    84 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 60,50 €
    60,50 €
    1 * 10 ST
       60,50 €
  • 1000 ml
    148 mm
    99 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 51,70 €
    51,70 €
    1 * 5 ST
       51,70 €
  • 2000 ml
    188 mm
    125 mm
    200 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,50 €
    16,50 €
    1 * 1 ST
       16,50 €