ContactUsLinkComponent
Beakers, tall form
Beakers, tall form
Catalog # 213-0036
Supplier:  SciLabware
Supplier #: 1015/18D
undefined
Beakers, tall form
Catalog # 213-0036
Supplier:  SciLabware
Supplier #: 1015/18D
Log in for availability
Select an Option
80 mm
Select an Option
600 ml
Select an Option
150 mm
Select an Option
10
About this item
PYREX®, borosilicate glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Volume graduations and labelling area.
Certifications: DIN 12331, ISO 3819
Frequently Bought Together