Orders LinkContactUsLinkComponent
Beakers, Philips
Beakers, Philips
Supplier:   SciLabware
Beakers, Philips
Supplier:  SciLabware
Certificates
Beakers, Philips
Beakers, Philips
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Conical shape with spout and labelling area.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    105 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 128,00 €
    128,00 €
    1 * 10 ST
       128,00 €
  • 500 ml
    142 mm
    86 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 242,00 €
    242,00 €
    1 * 10 ST
       242,00 €