ContactUsLinkComponent
VWR®, Beakers, Tall Form
  213-1169
 :
undefined
VWR®, Beakers, Tall Form
  213-1169
 :
Select an Option
38 mm
Select an Option
50 ml
Select an Option
70 mm
Select an Option
01
Select an Option
10
Borosilicate 3.3 glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
  • Volume graduations and labelling area
The economical alternative for a wide range of laboratory applications.
DIN 12331, ISO 3819
 : DIN 12331, ISO 3819
Frequently Bought Together