Orders LinkContactUsLinkComponent
VWR®, Beakers, Low Form
Supplier:   VWR Collection
VWR®, Beakers, Low Form
Supplier:  VWR Collection
Certificates
VWR®, Beakers, Low Form
VWR®, Beakers, Low Form
14 Options Available Below
Product Details & Documents
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 1 ml
    18 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 26,30 €
    26,30 €
    1 * 2 ST
       26,30 €
  • 5 ml
    26 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • 10 ml
    33 mm
    24 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,40 €
    21,40 €
    1 * 1 ST
       21,40 €
  • 25 ml
    47 mm
    32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,50 €
    24,50 €
    1 * 1 ST
       24,50 €
  • 50 ml
    60 mm
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,40 €
    31,40 €
    1 * 1 ST
       31,40 €
  • 100 ml
    68 mm
    54 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,20 €
    37,20 €
    1 * 1 ST
       37,20 €
  • 150 ml
    69 mm
    59 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,50 €
    48,50 €
    1 * 1 ST
       48,50 €
  • 250 ml
    97 mm
    66 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,80 €
    54,80 €
    1 * 1 ST
       54,80 €
  • 400 ml
    106 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,10 €
    82,10 €
    1 * 1 ST
       82,10 €
  • 500 ml
    125 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,20 €
    97,20 €
    1 * 1 ST
       97,20 €
  • 600 ml
    125 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
  • 1000 ml
    155 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • 2000 ml
    210 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 398,00 €
    398,00 €
    1 * 1 ST
       398,00 €
  • 5000 ml
    280 mm
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €