ContactUsLinkComponent
Beakers, tall form, DURAN®
  213-1161
 : 211165406
undefined
Beakers, tall form, DURAN®
  213-1161
 : 211165406
Select an Option
95 mm
Select an Option
1000 ml
Select an Option
180 mm
Select an Option
08
Select an Option
1
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
With printed retrace code (batch identification).
ISO 3819.
Frequently Bought Together