Orders LinkContactUsLinkComponent
Beakers, low form, Nalgene®
Beakers, low form, Nalgene®
   Sustainable Options Available
Beakers, low form, Nalgene®
   Sustainable Options Available
Certificates
Beakers, low form, Nalgene®
10 Options Available Below
Product Details & Documents
PPCO, translucent.
  • Very good chemical resistance
  • Heat resistant up to +121 °C, autoclavable at +121 °C (max. 20 minutes)
  • Smooth surfaces for easy, fast cleaning
Designed for general laboratory use. With spout and easy to read printed graduations with size code, plastic code, maximum temperature for use and 'Not flameproof' symbol. Ribbed for easy stacking. No drip, single spout design provides precision pouring control.
ISO 7056
Certifications: ISO 7056
Specifications for Product Family
Specifications
  • Catalog No:
  • 213-7100
  • 213-7101
  • 213-7102
  • 213-7103
  • 213-7104
  • 213-7105
  • 213-7106
  • 213-7107
  • 213-7108
  • 213-7109
  • Environmentally Preferable:
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Environmental certification:
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Graduation Range
    Availability
    Price
  • 30 ml
    5 - 30 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 45,50 €
    45,50 €
    1 * 12 ST
       45,50 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7100
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 50 ml
    10 - 50 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 49,10 €
    49,10 €
    1 * 12 ST
       49,10 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7101
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 100 ml
    10 - 100 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 53,90 €
    53,90 €
    1 * 12 ST
       53,90 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7102
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 150 ml
    20 - 50 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 61,60 €
    61,60 €
    1 * 12 ST
       61,60 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7103
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 250 ml
    25 - 250 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 43,00 €
    43,00 €
    1 * 6 ST
       43,00 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7104
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 400 ml
    50 - 400 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 55,50 €
    55,50 €
    1 * 6 ST
       55,50 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7105
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 600 ml
    100 - 600 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 47,80 €
    47,80 €
    1 * 4 ST
       47,80 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7106
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 1000 ml
    100 - 1000 ml
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 45,50 €
    45,50 €
    1 * 3 ST
       45,50 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7107
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 2000 ml
    200 - 2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,70 €
    32,70 €
    1 * 1 ST
       32,70 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7108
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 4000 ml
    500 - 4000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,80 €
    56,80 €
    1 * 1 ST
       56,80 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    213-7109
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Low Manufacturing Impact|Product Transparency and Disclosure