ContactUsLinkComponent
VWR®, Beakers, Low Form
VWR®, Beakers, Low Form
Catalog # 213-1122
Supplier:  VWR Collection
Supplier #:
undefined
VWR®, Beakers, Low Form
Catalog # 213-1122
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
50 mm
Select an Option
100 ml
Select an Option
70 mm
Select an Option
03
Select an Option
10
About this item
Borosilicate 3.3 glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
  • Volume graduations and labelling area
An economical alternative for a wide range of laboratory applications.
DIN 12331, ISO 3819
Certifications: DIN 12331, ISO 3819
Frequently Bought Together