Orders LinkContactUsLinkComponent
Wire baskets
Wire baskets
Supplier:   HUPFER METALLWERKE
Wire baskets
Supplier:  HUPFER METALLWERKE
Certificates
Wire baskets
9 Options Available Below
Product Details & Documents
Stainless steel.
  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges
These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 100×100×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • 120×80×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • 120×120×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 158,00 €
    158,00 €
    1 * 1 ST
       158,00 €
  • 140×140×140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 171,00 €
    171,00 €
    1 * 1 ST
       171,00 €
  • 160×160×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
  • 180×180×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
  • 200×200×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 211,00 €
    211,00 €
    1 * 1 ST
       211,00 €
  • 200×300×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 478,00 €
    478,00 €
    1 * 1 ST
       478,00 €
  • 200×400×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €