Bags, biohazard, Hi-Temp
Supplier: Alpha Packaging
Certificates
About this item
Bags with black warning symbol on yellow background. PP, 62,5 µm (250 gauge), transparent, with 'Biohazard' label
- Safety use instructions printed on each bag
- Suitable for temperatures up to 135 °C
Packaging: Supplied in benchtop dispenser boxes.
Frequently Bought Together