Order Entry
Austria
ContactUsLinkComponent
 
VWR®, Biohazard Bags
  129-0039
undefined
VWR®, Biohazard Bags
  129-0039

 

  • Capacity:
    12 L
  • L×W:
    610×305 mm
  • Material:
    HDPE
  • Seq:
    01
  • Pk:
    50

 

 

Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Supplied complete with the necessary sealing wires. HDPE, 100 µm thick, orange, with 'Biohazard' label.

  • Autoclavable up to 121 °C
  • Highly resistant to tears and impact
  • Luminous red for easy recognition and highly visible 'Biohazard' label with corresponding warning sign