Orders LinkContactUsLinkComponent
Adapters, delivery, angled | Avantor
Adapters, delivery, angled
Supplier:   Lenz Laborglas
Adapters, delivery, angled
Supplier:  Lenz Laborglas
Certificates
Adapters, delivery, angled
Adapters, delivery, angled
8 Options Available Below
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.
Dokumente
Product Family Options
Short tube
Product Information
  • Socket size
    Tube length
    Availability
    Price
  • 14/23 NS
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,40 €
    20,40 €
    1 * 1 ST
       20,40 €
  • 19/26 NS
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,50 €
    20,50 €
    1 * 1 ST
       20,50 €
  • 24/29 NS
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,00 €
    22,00 €
    1 * 1 ST
       22,00 €
  • 29/32 NS
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,00 €
    22,00 €
    1 * 1 ST
       22,00 €
Long tube
Product Information
  • Socket size
    Tube length
    Availability
    Price
  • 14/23 NS
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,60 €
    20,60 €
    1 * 1 ST
       20,60 €
  • 19/26 NS
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,80 €
    20,80 €
    1 * 1 ST
       20,80 €
  • 24/29 NS
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • 29/32 NS
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.