Orders LinkContactUsLinkComponent
Adapters, bends
Adapters, bends
Supplier:   Lenz Laborglas
Adapters, bends
Supplier:  Lenz Laborglas
Certificates
Adapters, bends
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 75°. With two standard ground cones.
Documents
Product Family Options
Product Information
  • Cone size
    Availability
    Price
  • 14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • 19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,30 €
    25,30 €
    1 * 1 ST
       25,30 €
  • 24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,20 €
    32,20 €
    1 * 1 ST
       32,20 €
  • 29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,50 €
    36,50 €
    1 * 1 ST
       36,50 €