Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
Supplier:   SciLabware
Multiple adapters, Quickfit®
Supplier:  SciLabware
Certificates
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
4 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,10 €
    81,10 €
    1 * 1 ST
       81,10 €
  • 24/29 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,00 €
    73,00 €
    1 * 1 ST
       73,00 €
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,10 €
    81,10 €
    1 * 1 ST
       81,10 €
  • 24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,20 €
    89,20 €
    1 * 1 ST
       89,20 €